4-Tolualdehyde ≥95.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: p-Tolualdehyde, p-Tolylaldehyde, p-Methylbenzaldehyd, 4-Methylbenzaldehyd, 4-Methylbenzaldehyde, 4-Tolylaldehyde
T1073-25ML T1073-500ML
TCIAT1073-25MLEA 17.6 EUR
TCIAT1073-25ML TCIAT1073-500ML
4-Tolualdehyde ≥95.0% (by GC)
4-Tolualdehyde
Formule: C₈H₈O
MW: 120.15 g/mol
Kookpunt: 204°C
Smeltpunt: -6
Vlampunt: 88°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00006954
CAS nummer: 104-87-0
EINECS: 203-246-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=644d421436d1a67a1c6eba54ca1cc6bfbfb98f3b&vid=01579efa68bea2364ac3b4ebcaad7a8a5ef1b88c&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 95.0 %