Benzylacetone ≥95.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: 4-Phenyl-2-butanone, 4-Phenylbutan-2-one
B0405-25ML B0405-500ML
TCIAB0405-25MLEA 17.6 EUR
TCIAB0405-25ML TCIAB0405-500ML
Benzylacetone ≥95.0% (by GC)
Benzylacetone
Formule: (C₆H₅)CH₂CH₂C(O)CH₃
MW: 148.2 g/mol
Kookpunt: 115°C
Smeltpunt: -13
Vlampunt: 98°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008790
CAS nummer: 2550-26-7
EINECS: 219-847-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1774cb7bf9a7e21599972fd2ac31503b676bc240&vid=6e863df7825f34ce4e8e5955930266d158c39424&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 95.0 %