4-Propylbenzoic acid ≥99.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

P0945-25G P0945-500G
TCIAP0945-25GEA 30.8 EUR
TCIAP0945-25G TCIAP0945-500G
4-Propylbenzoic acid ≥99.0% (by titrimetric analysis)
4-Propylbenzoic acid
Formule: C₁₀H₁₂O₂
Kookpunt: 298 °C (1013 hPa)
Smeltpunt: 142…144 °C
Dichtheid: ~ 1 g/cm³ (20 °C)
MDL Nummer: MFCD00013996
CAS nummer: 2438-05-3

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 99.0 %
Melting point 142.0 to 145.0 deg-C
Solubility in Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR