3-Amino-p-toluic acid ≥98.0% (by HPLC, titration analysis)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
A0920-25G A0920-250G
TCIAA0920-25GEA 31.9 EUR
TCIAA0920-25G TCIAA0920-250G
3-Amino-p-toluic acid ≥98.0% (by HPLC, titration analysis)
3-Amino-p-toluic acid
Formule: C₈H₉NO₂
Kookpunt: 363 °C (1013 hPa)
Smeltpunt: 166 °C
Dichtheid: 1 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00007742
CAS nummer: 2458-12-0
EINECS: 219-543-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ca9085eec59faef85a16ddd398a683a7894321fd&vid=964cad350d54149a97a10c1bc887d7c32a788f03&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White to Gray to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 165.0 to 169.0 deg-C
Solubility in hot Water almost transparency