α,2,4-Trichlorotoluene ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: 2,4-Dichlorobenzyl chloride, 1-(Chloromethyl)-2,4-dichlorobenzene, 1,3-Dichloro-4-(chloromethyl)benzene, 2,4-Dichloro-1-(chloromethyl)benzene
T0400-25G T0400-500G
TCIAT0400-25GEA 17.6 EUR
TCIAT0400-25G TCIAT0400-500G
α,2,4-Trichlorotoluene ≥98.0% (by GC)
α,2,4-Trichlorotoluene
Formule: C₇H₅Cl₃
MW: 195.47 g/mol
Kookpunt: 248°C
Smeltpunt: -2.7
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00000895
CAS nummer: 94-99-5
EINECS: 202-381-0
UN: 3265
ADR: 8 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cb1afe2be33d43d5a6488d9749c985c2097396ed&vid=2d0e268bc4f52b02955d674fe52123aa55c2b237&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %