3-Chloortolueen ≥99.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
C0296-25G C0296-500G
TCIAC0296-25GEA 27.5 EUR
TCIAC0296-25G TCIAC0296-500G
3-Chloortolueen ≥99.0% (by GC)
3-Chloortolueen
Formule: C₇H₇Cl
Kookpunt: 162°C
Smeltpunt: –48°C
Dichtheid: 1,0722 g/cm³ (20 °C)
Vlampunt: 53°C
MDL Nummer: MFCD00000595
CAS nummer: 108-41-8
EINECS: 203-580-5
UN: 2238
ADR: 3,III
Merck Index: 13,02190
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f70faaa421e63b09913f41ce635862fcf4201d2b&vid=1b985ee978fc9d9061e3ed0b3ec2fd69ae937ddb&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %