3-Chloro-α,α,α-trifluorotoluene ≥90.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: 1-Chloro-3-(trifluoromethyl)benzene, 3-Chlorobenzotrifluoride
C0304-25G C0304-500G
TCIAC0304-25GEA 18.7 EUR
TCIAC0304-25G TCIAC0304-500G
3-Chloro-α,α,α-trifluorotoluene ≥90.0% (by GC)
3-Chloro-α,α,α-trifluorotoluene
Formule: C₇H₄ClF₃
MW: 180.56 g/mol
Kookpunt: 139°C
Smeltpunt: -56
Vlampunt: 36°C
MDL Nummer: MFCD00000594
CAS nummer: 98-15-7
EINECS: 202-642-9
UN: 2234
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=33a183d1ec5be63dca5c0047a8c4c0744694db5f&vid=8959dabb6d05c5fdf41c90e6aa46ca9a900b50df&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 90.0 %