4-Chloro-2-fluorotoluene ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

C1513-25G C1513-250G
TCIAC1513-25GEA 29.7 EUR
TCIAC1513-25G TCIAC1513-250G
4-Chloro-2-fluorotoluene ≥98.0% (by GC)
4-Chloro-2-fluorotoluene
Formule: C₇H₆ClF
MW: 144.58 g/mol
Kookpunt: 153
Dichtheid: 1.184
Vlampunt: 51°C
MDL Nummer: MFCD00000571
CAS nummer: 452-75-5
EINECS: 207-210-3
UN: 1993
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e019b1c911e75fa8485e12d0d84a209165f3df33&vid=5f31f79808258c411f9b1c42a27993942d764b44&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR