α-Chloro-4-nitrotoluene ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
C0229-25G
C0229-250G
TCIAC0229-25GEA
36.3
EUR
TCIAC0229-25G
TCIAC0229-250G
α-Chloro-4-nitrotoluene ≥98.0% (by GC)
α-Chloro-4-nitrotoluene
Formule:
C₇H₆ClNO₂ Kookpunt: 293 °C (1013 hPa) Smeltpunt: 68…70 °C Dichtheid: 1,5647 g/cm³ (20 °C) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00007374 CAS nummer: 100-14-1 EINECS: 202-822-7 UN: 3261 ADR: 8,II |
|
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 70.0 to 75.0 deg-C |
Solubility in hot Methanol | faint turbidity |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...