3-Fluoro-4-nitrotoluene ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
F0567-25G
F0567-5G
TCIAF0567-25GEA
229
EUR
TCIAF0567-25G
TCIAF0567-5G
3-Fluoro-4-nitrotoluene ≥98.0% (by GC)
3-Fluoro-4-nitrotoluene
Formule:
C₇H₆FNO₂ Kookpunt: 98°C Smeltpunt: 53 °C Dichtheid: 1,438 g/cm³ (25 °C) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00007053 CAS nummer: 446-34-4 EINECS: 207-166-5 ADR: 6.1,III |
|
Specification Test Results
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 52.0 to 56.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...