α,α,α-Trifluoro-3,5-dinitrotoluene ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
D2699-25G D2699-5G
TCIAD2699-25GEA 70.4 EUR
TCIAD2699-25G TCIAD2699-5G
α,α,α-Trifluoro-3,5-dinitrotoluene ≥98.0% (by GC)
α,α,α-Trifluoro-3,5-dinitrotoluene
Formule: C₇H₃F₃N₂O₄
Kookpunt: 127 - 129 °C (9 -rr)
Smeltpunt: 50 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00007233
CAS nummer: 401-99-0
EINECS: 206-935-2
Order Now
Specification Test Results
Appearance White to Amber powder to crystal
Purity (GC) min. 98.0 %
Melting point 47.0 to 51.0 deg-C