Benzylamine ≥99.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
B0406-25ML B0406-500ML
TCIAB0406-25MLEA 18.7 EUR
TCIAB0406-25ML TCIAB0406-500ML
Benzylamine ≥99.0% (by GC)
Benzylamine
Formule: (C₆H₅)CH₂NH₂
Kookpunt: 184°C
Smeltpunt: 10 °C
Dichtheid: 0,98 g/cm³ (20 °C)
Vlampunt: 60°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008106
CAS nummer: 100-46-9
EINECS: 202-854-1
UN: 2734
ADR: 8,II
Merck Index: 14,01125
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7d966ec0c434d78128a66ba5bb2532b683b255d6&vid=5a2ef831079114cc46ed36ba351e99971aa362be&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %