4-tert-Butylphenol ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
B0230-25G B0230-500G
TCIAB0230-25GEA 17.6 EUR
TCIAB0230-25G TCIAB0230-500G
4-tert-Butylphenol ≥98.0% (by GC)
4-tert-Butylphenol
Formule: C₁₀H₁₄O
MW: 150.22 g/mol
Kookpunt: 237
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002367
CAS nummer: 98-54-4
EINECS: 202-679-0
UN: 2430
ADR: 8 III
Merck Index: 14,01585
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=217ab529ca3be5be7096bc6eb1373e3ab14503f8&vid=007787e1fd4bbe9d5eb43d0052e33f151f4ce3c9&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 98.0 to 101.0 deg-C
Solubility in Methanol almost transparency