4-Ethylphenol ≥97.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
E0159-25G E0159-500G
TCIAE0159-25GEA 17.6 EUR
TCIAE0159-25G TCIAE0159-500G
4-Ethylphenol ≥97.0% (by GC)
4-Ethylphenol
Formule: C₈H₁₀O
Kookpunt: 218 °C (1013 hPa)
Smeltpunt: 40…43 °C
Dichtheid: 1,011 g/cm³ (20 °C)
Vlampunt: 100 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002393
CAS nummer: 123-07-9
EINECS: 204-598-6
UN: 2430
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2e94a82ebb99c0fb59f2ef6cd90a2760cd11cc81&vid=f772e9bdb4eb2850b0110ba6f55af30b4fcc764d&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 97.0 %
Solubility in Methanol almost transparency