tert-Butylbenzene ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
B0715-25ML
B0715-500ML
TCIAB0715-25MLEA
17.6
EUR
TCIAB0715-25ML
TCIAB0715-500ML
tert-Butylbenzene ≥98.0% (by GC)
tert-Butylbenzene
Formule:
C₁₀H₁₄ MW: 134.22 g/mol Kookpunt: 168°C Smeltpunt: -58 Vlampunt: 60°C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00008816 CAS nummer: 98-06-6 EINECS: 202-632-4 UN: 2709 ADR: 3 III Merck Index: 14,01551 |
|
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...