Heptylbenzene ≥97.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: N-heptylbenzene, 1-Phenylheptane

H0046-25ML H0046-500ML
TCIAH0046-25MLEA 66 EUR
TCIAH0046-25ML TCIAH0046-500ML
Heptylbenzene ≥97.0% (by GC)
Heptylbenzene
Formule: C₁₃H₂₀
Kookpunt: 235°C
Smeltpunt: 83 °C
Dichtheid: 0,86 g/cm³ (20 °C)
Vlampunt: 95°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00009545
CAS nummer: 1078-71-3
EINECS: 214-084-3
UN: 3082
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8edf4d071576b2e90cf9e61b304b29da5de0edd2&vid=76186fe99abf320c5583952ecc01c0e9c69504e7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR