4-Methylbiphenyl ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M0563-25G
M0563-5G
TCIAM0563-25GEA
45.1
EUR
TCIAM0563-25G
TCIAM0563-5G
4-Methylbiphenyl ≥98.0% (by GC)
4-Methylbiphenyl
Formule:
C₁₃H₁₂ Kookpunt: 268°C Smeltpunt: 47…48 °C Dichtheid: 1,015 g/cm³ (27 °C) |
MDL Nummer:
MFCD00008544 CAS nummer: 644-08-6 EINECS: 211-409-0 |
|
Specification Test Results
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 46.0 to 49.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...