α-Bromo-o-xylene ≥97.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

B1004-25G B1004-100G
TCIAB1004-25GEA 91.3 EUR
TCIAB1004-25G TCIAB1004-100G
α-Bromo-o-xylene ≥97.0% (by GC)
α-Bromo-o-xylene
Formule: C₈H₉Br
Kookpunt: 216 °C (1013 hPa)
Smeltpunt: 18…20 °C
Dichtheid: 1,381 g/cm³ (20 °C)
Vlampunt: 82°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00000174
CAS nummer: 89-92-9
EINECS: 201-951-6
UN: 1701
ADR: 6.1,II
Merck Index: 12,10223
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e8c0eb020a349864f8a3267882af67cfb962ebce&vid=7b1596b30e91d9d758065d31552bcf53dc1e47bc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR