3-Bromo-o-xylene ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: 1-Bromo-2,3-dimethylbenzene
B1282-25G B1282-5G
TCIAB1282-25GEA 72.6 EUR
TCIAB1282-25G TCIAB1282-5G
3-Bromo-o-xylene ≥98.0% (by GC)
3-Bromo-o-xylene
Formule: C₈H₉Br
MW: 185.06 g/mol
Kookpunt: 214°C
Vlampunt: 80°C
MDL Nummer: MFCD00000069
CAS nummer: 576-23-8
EINECS: 209-398-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cb9ff111ad35e965b979f221d1125437d8c5ddc6&vid=715bf229a05d755b243c039e01eff3e66188f51a&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %