2-Bromo-p-xylene ≥99.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 2-Bromo-1,4-dimethylbenzene, 2,5-Dimethylbromobenzene

B0997-25G B0997-500G
TCIAB0997-25GEA 17.6 EUR
TCIAB0997-25G TCIAB0997-500G
2-Bromo-p-xylene ≥99.0% (by GC)
2-Bromo-p-xylene
Formule: C₈H₉Br
Kookpunt: 201°C
Dichtheid: 1,3582 g/cm³ (18.5 °C)
Vlampunt: 79°C
MDL Nummer: MFCD00000074
CAS nummer: 553-94-6
EINECS: 209-055-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ec0971bdcf0e90d8d6e45a6b1d4ce4ff9fde9831&vid=7423cee95973a0e59888b14025816e35d5f3ae39&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR