α'-Bromo-α,α,α-trifluoro-p-xylene ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
4-(Bromomethyl)benzotrifluoride, 1-(Bromomethyl)-4-(trifluoromethyl)benzene, 4-(Trifluoromethyl)benzyl bromide
T1313-25G
T1313-5G
TCIAT1313-25GEA
123
EUR
TCIAT1313-25G
TCIAT1313-5G
α'-Bromo-α,α,α-trifluoro-p-xylene ≥98.0% (by GC)
α'-Bromo-α,α,α-trifluoro-p-xylene
Formule:
C₈H₆BrF₃ MW: 239.03 g/mol Kookpunt: 66°C Smeltpunt: 31.5 Dichtheid: 1.54 Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00000403 CAS nummer: 402-49-3 EINECS: 206-947-8 UN: 3261 ADR: 8 II |
|
Specification Test Results
Appearance | White or Colorless to Light yellow powder to lump to clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...