3,5-Dimethylbenzaldehyde ≥96.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
D3498-5G D3498-25G
TCIAD3498-5GEA 51.7 EUR
TCIAD3498-5G TCIAD3498-25G
3,5-Dimethylbenzaldehyde ≥96.0% (by GC)
3,5-Dimethylbenzaldehyde
Formule: C₉H₁₀O
Kookpunt: 108°C
Smeltpunt: 9°C
Dichtheid: 1,09 g/cm³ (20 °C)
Vlampunt: 99°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00082777
CAS nummer: 5779-95-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4c1c12c240fab31cad6f3cbf26c42f12a92350ae&vid=28f8dae4f226e7d3e122617a65aacc48134250e3&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 96.0 %