Dichloro(diphenyl)silane ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
D0362-25G
D0362-500G
D0362-100G
TCIAD0362-25GEA
23.1
EUR
TCIAD0362-25G
TCIAD0362-500G
TCIAD0362-100G
Dichloro(diphenyl)silane ≥98.0% (by GC)
Dichloro(diphenyl)silane
Formule:
C₁₂H₁₀Cl₂Si Vlampunt: 157°C |
MDL Nummer:
MFCD00000489 CAS nummer: 80-10-4 EINECS: 201-251-0 UN: 1769 ADR: 8,II |
|
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...