tert-Butylnitriet ≥90.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
N0357-25ML N0357-250ML
TCIAN0357-25MLEA 45.1 EUR
TCIAN0357-25ML TCIAN0357-250ML
tert-Butylnitriet ≥90.0% (by GC)
tert-Butylnitriet
Formule: C₄H₉NO₂
Kookpunt: 63°C
Vlampunt: –13°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00002055
CAS nummer: 540-80-7
EINECS: 208-757-0
UN: 2351
ADR: 3,II
Merck Index: 14,01583
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c7c157208a8d671a7328589e7a4a383e70815578&vid=c7c157208a8d671a7328589e7a4a383e70815578&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Light yellow to Yellow clear liquid
Purity (GC) min. 90.0 %