4-Fluorophenethyl bromide ≥95.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: 1-Fluoro-4-(2-bromoethyl)benzene, 1-(2-bromoethyl)-4-fluorobenzene
F0431-1G F0431-5G
TCIAF0431-1GEA 23.1 EUR
TCIAF0431-1G TCIAF0431-5G
4-Fluorophenethyl bromide ≥95.0% (by GC)
4-Fluorophenethyl bromide
Formule: C₈H₈BrF
Kookpunt: 102°C
Vlampunt: 93°C
MDL Nummer: MFCD03844794
CAS nummer: 332-42-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=711bee5bcf4c877aa79178ec6efe356b6419ddab&vid=711bee5bcf4c877aa79178ec6efe356b6419ddab&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 95.0 %