4-Formylmorpholine ≥99.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

F0157-25G F0157-500G
TCIAF0157-25GEA 17.6 EUR
TCIAF0157-25G TCIAF0157-500G
4-Formylmorpholine ≥99.0% (by GC)
4-Formylmorpholine
Formule: C₅H₉NO₂
MDL Nummer: MFCD00006170
CAS nummer: 4394-85-8
EINECS: 224-518-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4547bee7492687e6042a205fba588494d3dca5de&vid=4547bee7492687e6042a205fba588494d3dca5de&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White or Colorless to Almost white or Almost colorless powder to lump to clear liquid
Purity (GC) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR