L(+)-Citrulline ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: H-Cit-OH

C0372-25G C0372-250G
TCIAC0372-25GEA 27.5 EUR
TCIAC0372-25G TCIAC0372-250G
L(+)-Citrulline ≥98.0% (by titrimetric analysis)
L(+)-Citrulline
Formule: C₆H₁₃N₃O₃
MW: 175.19 g/mol
Smeltpunt: 232°C
MDL Nummer: MFCD00064397
CAS nummer: 372-75-8
EINECS: 206-759-6
Merck Index: 14,02331
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6868b177618982b929ec6ff5041a43fca8a656e1&vid=6868b177618982b929ec6ff5041a43fca8a656e1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D +25.0 to +27.0 deg (C=8, 6mol/L HCl)
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR