Metanil yellow
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Acid Yellow 36, 3-(4-Anilinophenylazo)benzenesulfonic acid sodium salt
M0490-25G M0490-500G
TCIAM0490-25GEA 19.8 EUR
TCIAM0490-25G TCIAM0490-500G
Metanil yellow
Metanil yellow
Formule: C₁₈H₁₄N₃NaO₃S
MW: 375.38 g/mol
MDL Nummer: MFCD00007486
CAS nummer: 587-98-4
EINECS: 209-608-2
Merck Index: 14,05928
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cc299578c6a67921e4ffc554f9935f799c962f11&vid=cc299578c6a67921e4ffc554f9935f799c962f11&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Light yellow to Brown powder to crystal
Lambda max. 415.0 to 419.0 nm (MeOH)
Absorbance (E1%1cm) min. 500 (MeOH, 415.0 to 419.0 nm)