2,6-Dimethylbenzoic acid ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

D2260-25G D2260-5G
TCIAD2260-25GEA 250 EUR
TCIAD2260-25G TCIAD2260-5G
2,6-Dimethylbenzoic acid ≥98.0% (by GC, titration analysis)
2,6-Dimethylbenzoic acid
Formule: C₉H₁₀O₂
Kookpunt: 160°C
Smeltpunt: 115…116 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002483
CAS nummer: 632-46-2
EINECS: 211-177-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f3e85804e9dac4c158d26b993206d9d5b5b6f7b4&vid=437a629fab2a6371c692167c7728bff2777ebfeb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 115.0 to 117.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR