3,4-Dimethylbenzoic acid ≥99.0% (by titrimetric analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
D1439-10G
D1439-25G
TCIAD1439-10GEA
26.4
EUR
TCIAD1439-10G
TCIAD1439-25G
3,4-Dimethylbenzoic acid ≥99.0% (by titrimetric analysis)
3,4-Dimethylbenzoic acid
Formule:
C₉H₁₀O₂ Kookpunt: 295 °C (1013 hPa) Smeltpunt: 163…164 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00002524 CAS nummer: 619-04-5 EINECS: 210-576-7 |
|
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 99.0 % |
Melting point | 165.0 to 169.0 deg-C |
Solubility in Methanol | very faint turbidity |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...