3,5-Dimethylbenzoic acid ≥99.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

D0684-25G D0684-250G
TCIAD0684-25GEA 52.8 EUR
TCIAD0684-25G TCIAD0684-250G
3,5-Dimethylbenzoic acid ≥99.0% (by titrimetric analysis)
3,5-Dimethylbenzoic acid
Formule: C₉H₁₀O₂
Kookpunt: 278 °C (1013 hPa)
Smeltpunt: 166…169 °C
Dichtheid: 1,211 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002525
CAS nummer: 499-06-9
EINECS: 207-876-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7d793f80c24849e07cf74f3ff7cd6ce35cbea4b9&vid=ce108e06f4644bf5e99c2208a9a6b51c51c0c930&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (Neutralization titration) min. 99.0 %
Melting point 168.0 to 172.0 deg-C
Solubility in Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR