α,α'-Dichloro-p-xylene ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

Synoniemen: 1,4-Bis(chloromethyl)benzene, p-Xylene dichloride, p-Xylylene dichloride

D0430-25G D0430-500G D0430-100G
TCIAD0430-25GEA 19.8 EUR
TCIAD0430-25G TCIAD0430-500G TCIAD0430-100G
α,α'-Dichloro-p-xylene ≥98.0% (by GC)
α,α'-Dichloro-p-xylene
Formule: C₈H₈Cl₂
MW: 175.06 g/mol
Kookpunt: 142°C
Smeltpunt: 100
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00000920
CAS nummer: 623-25-6
EINECS: 210-782-7
UN: 2928
ADR: 6.1 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eea1d845c0e08a85b1153acac001db86a699ad75&vid=e2c77c6fc87062050b9cdf9059638c30b948c6e9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 100.0 to 102.0 deg-C
Solubility in Toluene almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR