4-Iodo-m-xylene ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
I0561-25G I0561-5G
TCIAI0561-25GEA 70.4 EUR
TCIAI0561-25G TCIAI0561-5G
4-Iodo-m-xylene ≥98.0% (by GC)
4-Iodo-m-xylene
Formule: C₈H₉I
Kookpunt: 233°C
Dichtheid: 1,6609 g/cm³ (13 °C)
MDL Nummer: MFCD00013706
CAS nummer: 4214-28-2
EINECS: 224-141-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=263c07852d01ae51b40e86998d5565817f1c2ce4&vid=624b9737eb3aaef2d57a2bf420d6f2bc1ad26701&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Brown clear liquid
Purity (GC) min. 98.0 %