2,6-Xylenol ≥99.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
2,6-Dimethylphenol, 2-Hydroxy-m-xylene, 2-Hydroxy-3-xylene
D0776-25G
D0776-500G
TCIAD0776-25GEA
19.8
EUR
TCIAD0776-25G
TCIAD0776-500G
2,6-Xylenol ≥99.0% (by GC)
2,6-Xylenol
Formule:
C₈H₁₀O MW: 122.17 g/mol Kookpunt: 203 Vlampunt: 73 Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00002240 CAS nummer: 576-26-1 EINECS: 209-400-1 UN: 2261 ADR: 6.1 II Merck Index: 14,10082 |
|
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 99.0 % |
Melting point | 45.0 to 48.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...