2,5-Xylenol ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
D0775-25G D0775-500G D0775-100G
TCIAD0775-25GEA 18.7 EUR
TCIAD0775-25G TCIAD0775-500G TCIAD0775-100G
2,5-Xylenol ≥98.0% (by GC)
2,5-Xylenol
Formule: C₈H₁₀O
Kookpunt: 212 °C (1013 hPa)
Smeltpunt: 75 °C
Dichtheid: 1,169 g/cm³ (15 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002237
CAS nummer: 95-87-4
EINECS: 202-461-5
UN: 2261
ADR: 6.1,II
Merck Index: 14,10082
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6d0f60b8c1b26e00a00e08e113fe0c7c40388c12&vid=6c6ad57f4aaf49f490bc8a357e9be16282be5f1b&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 75.0 to 77.0 deg-C
Solubility in Methanol almost transparency