2,4-Dimethylthiophenol ≥96.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: 2,4-Dimethylbenzene-1-thiol, 1,3-Dimethyl-4-mercaptobenzene, 4-Mercapto-m-xylene, 2,4-Dimethylbenzenethiol
D2648-25G D2648-5G
TCIAD2648-25GEA 120 EUR
TCIAD2648-25G TCIAD2648-5G
2,4-Dimethylthiophenol ≥96.0% (by GC)
2,4-Dimethylthiophenol
Formule: C₈H₁₀S
Kookpunt: 214°C
Dichtheid: 1,027 g/cm³ (20 °C)
Vlampunt: 58°C
MDL Nummer: MFCD00010019
CAS nummer: 13616-82-5
EINECS: 237-100-0
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a8fcd4f30d31defaed597306c94b3901a4e4795e&vid=a1c8c19502eff66c24795536796ae5e8537787d6&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 96.0 %