2-Bromomesitylene ≥99.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
2,4,6-Trimethylbromobenzene, 2-Bromo-1,3,5-trimethylbenzene, Mesityl bromide, 2-Bromomesitylene
B1261-25G
B1261-500G
B1261-100G
TCIAB1261-25GEA
25.3
EUR
TCIAB1261-25G
TCIAB1261-500G
TCIAB1261-100G
2-Bromomesitylene ≥99.0% (by GC)
2-Bromomesitylene
Formule:
C₉H₁₁Br Kookpunt: 225°C Smeltpunt: 2 °C Dichtheid: 1,32 g/cm³ (20 °C) Vlampunt: 96°C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00000073 CAS nummer: 576-83-0 EINECS: 209-405-9 |
|
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...