2,4,6-Trimethylaniline ≥99.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
T0467-25ML T0467-500ML T0467-100ML
TCIAT0467-25MLEA 17.6 EUR
TCIAT0467-25ML TCIAT0467-500ML TCIAT0467-100ML
2,4,6-Trimethylaniline ≥99.0% (by GC)
2,4,6-Trimethylaniline
Formule: C₉H₁₃N
MW: 135.21 g/mol
Kookpunt: 230°C
Smeltpunt: -5
Vlampunt: 96°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00007740
CAS nummer: 88-05-1
EINECS: 201-794-3
UN: 2810
ADR: 6.1 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=25ae60899f7fe1c1d68ae87c386c965f87671558&vid=2f9157c6ca25c394db1998b3d9aee90ffee937a4&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Yellow to Orange clear liquid
Purity (GC) min. 99.0 %
Purity (Nonaqueous Titration) min. 98.0 %