2,3,5-Trimethylphenol ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
T0485-25G T0485-500G
TCIAT0485-25GEA 38.5 EUR
TCIAT0485-25G TCIAT0485-500G
2,3,5-Trimethylphenol ≥98.0% (by GC)
2,3,5-Trimethylphenol
Formule: C₉H₁₂O
Kookpunt: 233°C
Smeltpunt: 92 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002228
CAS nummer: 697-82-5
EINECS: 211-806-9
UN: 2430
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=243387864fa3633b9bca3cc57430c7c5fa508e24&vid=4e45bc1b55d59341d575a0282f05fd75fb9992d5&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 93.0 to 97.0 deg-C