1,3,5-Tri-tert-butylbenzene ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
T1633-25G T1633-5G
TCIAT1633-25GEA 83.6 EUR
TCIAT1633-25G TCIAT1633-5G
1,3,5-Tri-tert-butylbenzene ≥98.0% (by GC)
1,3,5-Tri-tert-butylbenzene
Formule: C₁₈H₃₀
MW: 246.44 g/mol
Kookpunt: 248°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008831
CAS nummer: 1460-02-2
EINECS: 215-952-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ec9fce5e8d19e9a81fe1a52b5544ef169800f0ec&vid=4f9350d75b12a135106b7df856fccb60922605cb&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 71.0 to 75.0 deg-C