1,3,5-Triethylbenzeen ≥97.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
T0426-25ML
T0426-5ML
TCIAT0426-25MLEA
515
EUR
TCIAT0426-25ML
TCIAT0426-5ML
1,3,5-Triethylbenzeen ≥97.0% (by GC)
1,3,5-Triethylbenzeen
Formule:
C₁₂H₁₈ Kookpunt: 218°C Smeltpunt: 3 °C Dichtheid: 0,8631 g/cm³ (20 °C) Vlampunt: 76°C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00009261 CAS nummer: 102-25-0 EINECS: 203-017-3 UN: 3082 ADR: 9,III |
|
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...