1-Phenylcyclohexanol ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

P1988-5G P1988-25G
TCIAP1988-5GEA 53.9 EUR
TCIAP1988-5G TCIAP1988-25G
1-Phenylcyclohexanol ≥98.0% (by GC)
1-Phenylcyclohexanol
Formule: C₁₂H₁₆O
MW: 176.26 g/mol
Kookpunt: 100°C
Dichtheid: 1.101
MDL Nummer: MFCD00021393
CAS nummer: 1589-60-2
EINECS: 216-456-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a5ebf88b851149fabadc0597e2a853476d3de197&vid=89900879c1a763013b2f8fac098fd897daf3893b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 61.0 to 65.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR