2-Phenylbenzoic acid ≥98.0% (by GC, titration analysis)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
P0906-100G P0906-25G P0906-500G
TCIAP0906-100GEA 208 EUR
TCIAP0906-100G TCIAP0906-25G TCIAP0906-500G
2-Phenylbenzoic acid ≥98.0% (by GC, titration analysis)
2-Phenylbenzoic acid
Formule: C₁₃H₁₀O₂
Kookpunt: 344°C
Smeltpunt: 108…109 °C
Dichtheid: 1,458 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002463
CAS nummer: 947-84-2
EINECS: 213-432-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ba5b90ef373641d1cedfbd0e770c8f9bfaa82685&vid=885a9e355d7d2ce4d9e5ebe047d053484531ddfe&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White to Light yellow to Light orange powder to lump
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 111.0 to 114.0 deg-C
Solubility in Methanol very faint turbidity