4-Quinolinol ≥98.0% (by GC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Q0029-25G
Q0029-5G
TCIAQ0029-25GEA
286
EUR
TCIAQ0029-25G
TCIAQ0029-5G
4-Quinolinol ≥98.0% (by GC, titration analysis)
4-Quinolinol
Formule:
C₉H₇NO Smeltpunt: 202…206 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00006777 CAS nummer: 611-36-9 EINECS: 210-268-2 |
|
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 202.0 to 206.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...