8-Nitroquinaldine ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 2-Methyl-8-nitroquinoline

N0539-25G N0539-5G
TCIAN0539-25GEA 226 EUR
TCIAN0539-25G TCIAN0539-5G
8-Nitroquinaldine ≥98.0% (by GC, titration analysis)
8-Nitroquinaldine
Formule: C₁₀H₈N₂O₂
MDL Nummer: MFCD00006764
CAS nummer: 881-07-2
EINECS: 212-919-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe4b649c312d3b83b74081488aefcca1369cfd09&vid=a6828a8e6c23498c65b12219f5725162c6043a41&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 138.0 to 141.0 deg-C
Solubility in hot Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR