6-Methylquinaldine ≥99.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
D1236-25G D1236-5G
TCIAD1236-25GEA 560 EUR
TCIAD1236-25G TCIAD1236-5G
6-Methylquinaldine ≥99.0% (by GC)
6-Methylquinaldine
Formule: C₁₁H₁₁N
Kookpunt: 267°C
Smeltpunt: 55…58 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00006762
CAS nummer: 877-43-0
EINECS: 212-891-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=90d4b66dc6d60a0f076fe028d3dabce13bbb87e6&vid=41f1a96404e31410a9a5fa489bc54358ed3b8498&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Melting point 57.0 to 60.0 deg-C
Solubility in Methanol almost transparency