Bromocyclopentane ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
B1006-25G
B1006-500G
B1006-100G
TCIAB1006-25GEA
22
EUR
TCIAB1006-25G
TCIAB1006-500G
TCIAB1006-100G
Bromocyclopentane ≥98.0% (by GC)
Bromocyclopentane
Formule:
C₅H₉Br Kookpunt: 138°C Dichtheid: 1,39 g/cm³ (20 °C) Vlampunt: 35°C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00001359 CAS nummer: 137-43-9 EINECS: 205-294-6 UN: 1993 ADR: 3,III |
|
Specification Test Results
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...