1,3-Cyclopentanedione ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
Cyclopentane-1,3-dione
C1146-1G
C1146-5G
TCIAC1146-1GEA
34.1
EUR
TCIAC1146-1G
TCIAC1146-5G
1,3-Cyclopentanedione ≥98.0% (by GC)
1,3-Cyclopentanedione
Formule:
C₅H₆O₂ Smeltpunt: 149…152 °C Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00001405 CAS nummer: 3859-41-4 EINECS: 223-372-8 |
|
Specification Test Results
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...