Cyclohexanethiol ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: Mercaptocyclohexane, Cyclohexyl Mercaptan

C0727-25ML C0727-500ML
TCIAC0727-25MLEA 31.9 EUR
TCIAC0727-25ML TCIAC0727-500ML
Cyclohexanethiol ≥98.0% (by GC, titration analysis)
Cyclohexanethiol
Formule: C₆H₁₂S
Kookpunt: 158°C
Smeltpunt: –72 °C
Dichtheid: 0,95 g/cm³ (20 °C)
Vlampunt: 43°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00001455
CAS nummer: 1569-69-3
EINECS: 216-378-7
UN: 3054
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c57355e1fba24f75aaaf64697037900aea525533&vid=c57355e1fba24f75aaaf64697037900aea525533&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR