Cyclopentanethiol ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: Cyclopentyl mercaptan, Mercaptocyclopentane

C0810-25ML C0810-5ML
TCIAC0810-25MLEA 124 EUR
TCIAC0810-25ML TCIAC0810-5ML
Cyclopentanethiol ≥98.0% (by GC)
Cyclopentanethiol
Formule: C₅H₁₀S
Kookpunt: 132°C
Dichtheid: 0,957 g/cm³ (20 °C)
Vlampunt: 25°C
MDL Nummer: MFCD00001369
CAS nummer: 1679-07-8
EINECS: 216-841-3
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=67697de763a962c48e3cd7070d8a73e16a12a169&vid=67697de763a962c48e3cd7070d8a73e16a12a169&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR